siRNA Details
| Gene Symbol: Ubr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025666.2 | NP_079942.1 | 2 | 279 | ||
| GenBank mRNA | AK011968.1 | - | 2 | 150 | ||
| AK017542.1 | - | 2 | 234 | |||
| AK088208.1 | - | 2 | 281 | |||
| AK146308.1 | - | 2 | 280 | |||
| AK149989.1 | - | 2 | 234 | |||
| AK155611.1 | - | 2 | 150 | |||
| AK190790.1 | - | 1 | 13 | |||
| BC094323.1 | - | 2 | 287 | |||
| BC094423.1 | - | 2 | 237 | 6812389 | ||

Ubr7
Genomic Map