siRNA Details
| Gene Symbol: Mcm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291054.1 | NP_001277983.1 | 2 | 446 | ||
| NM_025676.4 | NP_079952.2 | 2 | 446 | |||
| GenBank mRNA | AK012845.1 | - | 2 | 405 | ||
| AK017607.1 | - | 2 | 344 | |||
| AK019268.1 | - | 2 | 405 | |||
| AK133858.1 | - | 2 | 403 | |||
| BC046780.1 | - | 2 | 393 | 5716136 | ||
| BC052070.1 | - | 2 | 252 | 5716125 | ||

Mcm8
Genomic Map