siRNA Details
| Gene Symbol: Rpe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025683.2 | NP_079959.2 | 3 | 281 | ||
| GenBank mRNA | AK007735.1 | - | 3 | 287 | ||
| AK017774.1 | - | 3 | 282 | |||
| AK029088.1 | - | 3 | 280 | |||
| AK088152.1 | - | 3 | 282 | |||
| AK150008.1 | - | 3 | 319 | |||
| BC006953.1 | - | 3 | 275 | |||
| BC019126.1 | - | 3 | 324 | 5008075 | ||
| Z78161.1 | - | 4 | 300 | |||

Rpe
Genomic Map