siRNA Details
| Gene Symbol: Spesp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025721.2 | NP_079997.1 | 2 | 278 | ||
| GenBank mRNA | AK014843.1 | - | 2 | 279 | ||
| AK015620.1 | - | 2 | 309 | |||
| BC050754.1 | - | 2 | 286 | 6773961 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spesp1| Gene Symbol: Spesp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025721.2 | NP_079997.1 | 2 | 278 | ||
| GenBank mRNA | AK014843.1 | - | 2 | 279 | ||
| AK015620.1 | - | 2 | 309 | |||
| BC050754.1 | - | 2 | 286 | 6773961 | ||