siRNA Details
| Gene Symbol: Lrrk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025730.3 | NP_080006.3 | 42 | 6224 | ||
| GenBank mRNA | AK014886.1 | - | 3 | 283 | ||
| AK014938.1 | - | 3 | 296 | |||
| AY792512.1 | - | 42 | 6167 | |||
| BC034074.1 | - | 8 | 1107 | |||

Lrrk2
Genomic Map