siRNA Details
| Gene Symbol: Erich2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025744.2 | NP_080020.2 | 3 | 841 | ||
| GenBank mRNA | AK006384.1 | - | 3 | 170 | ||
| AK016654.1 | - | 3 | 842 | |||
| AK017113.1 | - | 3 | 180 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Erich2| Gene Symbol: Erich2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025744.2 | NP_080020.2 | 3 | 841 | ||
| GenBank mRNA | AK006384.1 | - | 3 | 170 | ||
| AK016654.1 | - | 3 | 842 | |||
| AK017113.1 | - | 3 | 180 | |||