siRNA Details
| Gene Symbol: Oser1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025699.2 | NP_079975.2 | 4 | 1026 | ||
| GenBank mRNA | AK006969.1 | - | 4 | 1069 | ||
| AK009708.1 | - | 4 | 1042 | |||
| AK014323.1 | - | 4 | 1045 | |||
| AK088622.1 | - | 4 | 1045 | |||
| AK139692.1 | - | 1198 | ||||
| AK153499.1 | - | 4 | 1063 | |||
| BC005579.1 | - | 4 | 1026 | 3968876 | ||

Oser1
Genomic Map