siRNA Details
| Gene Symbol: Tmem186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025708.4 | NP_079984.2 | 2 | 2659 | ||
| GenBank mRNA | AK013859.1 | - | 1 | 157 | ||
| AK014478.1 | - | 2 | 2658 | |||
| AK088656.1 | - | 2 | 2669 | |||
| AK159727.1 | - | 2 | 2645 | |||
| AK159782.1 | - | 2 | 2645 | |||
| BC027261.1 | - | 2 | 2489 | 2649343 | ||

Tmem186
Genomic Map