siRNA Details
| Gene Symbol: Aspn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172481.1 | NP_001165952.1 | 4 | 844 | ||
| NM_025711.3 | NP_079987.2 | 4 | 861 | |||
| GenBank mRNA | AF316825.1 | - | 3 | 581 | ||
| AK014504.1 | - | 4 | 817 | |||
| AK085920.1 | - | 4 | 816 | |||
| BC034888.1 | - | 4 | 763 | 1365428 | ||
| BC145904.1 | - | 4 | 759 | 40131207 | ||
| BC145930.1 | - | 4 | 742 | 40131233 | ||

Aspn
Genomic Map