siRNA Details
| Gene Symbol: Rbm4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025717.3 | NP_079993.2 | 4 | 1350 | ||
| GenBank mRNA | AK142116.1 | - | 4 | 1306 | ||
| BC019488.1 | - | 4 | 1255 | 4457936 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbm4b| Gene Symbol: Rbm4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025717.3 | NP_079993.2 | 4 | 1350 | ||
| GenBank mRNA | AK142116.1 | - | 4 | 1306 | ||
| BC019488.1 | - | 4 | 1255 | 4457936 | ||