siRNA Details
| Gene Symbol: CPT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000098.2 | NP_000089.1 | 2, 3 | 756 | ||
| XM_005270484.1 | XP_005270541.1 | 2, 3 | 424 | |||
| GenBank mRNA | AK312687.1 | - | 2, 3 | 360 | ||
| BC002445.2 | - | 2, 3 | 365 | 3346025 | ||
| BC005172.2 | - | 2, 3 | 365 | 3605012 | ||
| M58581.1 | - | 2, 3 | 342 | |||
| U09648.1 | - | 2, 3 | 757 | |||

CPT2
Genomic Map