siRNA Details
| Gene Symbol: Erlec1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025745.3 | NP_080021.3 | 10, 11 | 1165 | ||
| GenBank mRNA | AK002471.1 | - | 11, 12 | 1156 | ||
| AK077120.1 | - | 10, 11 | 1158 | |||
| AK133874.1 | - | 10, 11 | 1166 | |||
| BC018468.1 | - | 10, 11 | 1175 | 3591901 | ||

Erlec1
Genomic Map