siRNA Details
| Gene Symbol: Cfap97 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025747.3 | NP_080023.1 | 1 | 157 | ||
| GenBank mRNA | AK016776.1 | - | 2 | 175 | ||
| AK020776.1 | - | 2 | 185 | |||
| AK020894.1 | - | 3 | 440 | |||
| AK048125.1 | - | 2 | 243 | |||
| AK082519.1 | - | 3 | 348 | |||
| AK129358.1 | - | 2 | 215 | |||
| BC049135.1 | - | 2 | 201 | 6831539 | ||
| BC058092.1 | - | 1 | 159 | 6821844 | ||

Cfap97
Genomic Map