siRNA Details
| Gene Symbol: Asb17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025758.4 | NP_080034.2 | 1 | 405 | ||
| GenBank mRNA | AF425641.1 | - | 1 | 405 | ||
| AK016273.1 | - | 1 | 405 | |||
| AK017090.1 | - | 1 | 387 | |||
| BC049572.1 | - | 1 | 410 | 6773716 | ||

Asb17
Genomic Map