siRNA Details
Gene Symbol: Speer4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025759.3 | NP_080035.2 | 4 | 633 | ||
GenBank mRNA | AK017002.1 | - | 4 | 634 |
Gene Symbol: Gm10354 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281514.1 | NP_001268443.1 | 4 | 623 |
Gene Symbol: Speer4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281511.1 | NP_001268440.1 | 4 | 668 |
