siRNA Details
| Gene Symbol: Aknaos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005960.1 | - | 355 | |||
| AK017104.1 | - | 901 | ||||
| AK018846.1 | - | 353 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aknaos| Gene Symbol: Aknaos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005960.1 | - | 355 | |||
| AK017104.1 | - | 901 | ||||
| AK018846.1 | - | 353 | ||||