siRNA Details
| Gene Symbol: Thap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025780.3 | NP_080056.1 | 1, 2 | 239 | ||
| GenBank mRNA | AK009162.1 | - | 1, 2 | 218 | ||
| AK018577.1 | - | 1, 2 | 219 | |||
| AK081303.1 | - | 1, 2 | 240 | |||
| BC027502.1 | - | 1, 2 | 147 | 1515752 | ||

Thap2
Genomic Map