siRNA Details
| Gene Symbol: Wdr45b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025793.3 | NP_080069.2 | 3 | 319 | ||
| GenBank mRNA | AK002352.1 | - | 3 | 280 | ||
| AK013170.1 | - | 3 | 269 | |||
| AK146101.1 | - | 3 | 320 | |||
| AK151975.1 | - | 3 | 281 | |||
| AK168603.1 | - | 3 | 272 | |||
| AK170602.1 | - | 3 | 266 | |||
| AK182151.1 | - | 167 | ||||
| AK188918.1 | - | 2 | 118 | |||
| AK189483.1 | - | 3 | 119 | |||
| AK215641.1 | - | 3 | 118 | |||
| BC004595.1 | - | 3 | 263 | 3584839 | ||

Wdr45b
Genomic Map