siRNA Details
| Gene Symbol: Etfdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025794.2 | NP_080070.2 | 2, 3 | 396 | ||
| GenBank mRNA | AK002483.1 | - | 2, 3 | 349 | ||
| AK028686.1 | - | 2, 3 | 348 | |||
| AK037969.1 | - | 2, 3 | 313 | |||
| AK075673.1 | - | 2, 3 | 347 | |||
| AK085493.1 | - | 2, 3 | 365 | |||
| AK141684.1 | - | 2, 3 | 350 | |||
| AK152624.1 | - | 2, 3 | 365 | |||
| AK169686.1 | - | 2, 3 | 301 | |||
| BC012522.1 | - | 2, 3 | 345 | 2648103 | ||
| BC057670.1 | - | 2, 3 | 363 | 2810976 | ||

Etfdh
Genomic Map