siRNA Details
| Gene Symbol: Lonp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025827.3 | NP_080103.1 | 2 | 409 | ||
| GenBank mRNA | AK004843.1 | - | 2 | 323 | ||
| AK050231.1 | - | 2 | 318 | |||
| AK088348.1 | - | 2 | 410 | |||
| AK151103.1 | - | 2 | 355 | |||
| AK168628.1 | - | 2 | 359 | |||
| AK168791.1 | - | 2 | 355 | |||
| AK168956.1 | - | 2 | 355 | |||
| AK169656.1 | - | 323 | ||||
| AK200906.1 | - | 2 | 121 | |||
| BC049090.1 | - | 2 | 345 | 6401542 | ||

Lonp2
Genomic Map