siRNA Details
| Gene Symbol: Vps28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF373710.1 | - | 3, 4 | 195 | ||
| AK003572.2 | - | 3, 4 | 225 | |||
| AK003699.1 | - | 3, 4 | 149 | |||
| AK085669.1 | - | 3, 4 | 274 | |||
| BC013535.1 | - | 3, 4 | 158 | 4239189 | ||
| BY083019.1 | - | 3, 4 | 207 | |||

Vps28
Genomic Map