siRNA Details
| Gene Symbol: Ndufb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025843.3 | NP_080119.1 | 3 | 369 | ||
| GenBank mRNA | AK002678.1 | - | 3 | 367 | ||
| AK003300.1 | - | 3 | 370 | |||
| BC034787.1 | - | 3 | 348 | 4913656 | ||
| BC056984.1 | - | 2 | 4210 | |||

Ndufb7
Genomic Map