siRNA Details
| Gene Symbol: Pqlc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164423.1 | NP_001157895.1 | 4 | 591 | ||
| NM_001164424.1 | NP_001157896.1 | 4 | 586 | |||
| NM_025861.3 | NP_080137.2 | 4 | 591 | |||
| GenBank mRNA | AK017854.1 | - | 4 | 599 | ||
| AK018759.1 | - | 3 | 969 | |||
| AK047446.1 | - | 4 | 524 | |||
| BC043686.1 | - | 769 | 5135928 | |||

Pqlc1
Genomic Map