siRNA Details
| Gene Symbol: Efcab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025769.3 | NP_080045.1 | 3 | 343 | ||
| GenBank mRNA | AK015426.1 | - | 325 | |||
| AK015866.1 | - | 3 | 363 | |||
| AK017267.1 | - | 3 | 344 | |||
| AK030381.1 | - | 3 | 353 | |||
| AK038735.1 | - | 3 | 353 | |||
| AK042748.1 | - | 3 | 385 | |||
| AK043519.1 | - | 3 | 315 | |||
| AK077722.1 | - | 3 | 346 | |||
| BC085167.1 | - | 3 | 235 | 30538702 | ||

Efcab1
Genomic Map