siRNA Details
| Gene Symbol: Ttc39b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027238.2 | NP_081514.1 | 20 | 6620 | ||
| GenBank mRNA | AK021142.1 | - | 1 | 757 | ||
| AK031930.1 | - | 1 | 2683 | |||
| AK134680.1 | - | 1 | 591 | |||
| BC020085.1 | - | 1 | 448 | |||
| BC151021.1 | - | 20 | 6593 | 9087934 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_025782.1 | - | 2683 | Chr. - - [-] on Build | ||

Ttc39b
Genomic Map