siRNA Details
| Gene Symbol: Fbxo25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025785.2 | NP_080061.1 | 3 | 270 | ||
| GenBank mRNA | AK008027.1 | - | 3 | 274 | ||
| AK018626.1 | - | 3 | 271 | |||
| AK134722.1 | - | 3 | 274 | |||
| BC014749.1 | - | 3 | 256 | 4240953 | ||

Fbxo25
Genomic Map