siRNA Details
| Gene Symbol: Cyb5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025797.3 | NP_080073.1 | 5 | 632 | ||
| GenBank mRNA | AK002426.1 | - | 5 | 633 | ||
| AK013210.1 | - | 5 | 633 | |||
| AK132102.1 | - | 5 | 655 | |||
| BC024341.1 | - | 5 | 637 | 4973059 | ||

Cyb5a
Genomic Map