siRNA Details
| Gene Symbol: Hmg20a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025812.2 | NP_080088.1 | 3 | 306 | ||
| GenBank mRNA | AK004596.1 | - | 3 | 326 | ||
| AK017716.1 | - | 3 | 509 | |||
| AK032790.1 | - | 3 | 334 | |||
| AK039222.1 | - | 3 | 328 | |||
| AK086346.1 | - | 3 | 324 | |||
| AK129010.1 | - | 3 | 508 | |||
| AK134213.1 | - | 3 | 478 | |||
| AK154075.1 | - | 3 | 325 | |||
| AL355705.1 | - | 3 | 292 | |||
| BC013804.1 | - | 3 | 306 | 3602239 | ||
| BC068257.1 | - | 3 | 379 | 6434284 | ||
| DQ182735.1 | - | 3 | 306 | |||

Hmg20a
Genomic Map