siRNA Details
| Gene Symbol: 1700010I14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025851.3 | NP_080127.2 | 2 | 192 | ||
| GenBank mRNA | AK005830.1 | - | 2 | 172 | ||
| AK006307.1 | - | 2 | 164 | |||
| AK015220.1 | - | 4 | 371 | |||
| AK161278.1 | - | 2 | 169 | |||
| BC050753.1 | - | 2 | 164 | |||
| BC061078.1 | - | 2 | 142 | 6770439 | ||

1700010I14Rik
Genomic Map