siRNA Details
| Gene Symbol: Ddx18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025860.3 | NP_080136.2 | 2 | 261 | ||
| GenBank mRNA | AK012706.1 | - | 2 | 229 | ||
| AK012878.1 | - | 2 | 201 | |||
| AK019845.1 | - | 2 | 185 | |||
| AK075864.1 | - | 2 | 198 | |||
| BC017160.1 | - | 2 | 183 | |||
| BC028246.1 | - | 2 | 232 | 2582151 | ||
| BC103776.1 | - | 2 | 198 | 6492142 | ||
| BC141230.1 | - | 2 | 200 | 9055842 | ||
| BC141231.1 | - | 2 | 200 | 9055843 | ||
| BY084565.1 | - | 2 | 262 | |||

Ddx18
Genomic Map