siRNA Details
| Gene Symbol: Acad8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025862.2 | NP_080138.2 | 2, 3 | 265 | ||
| GenBank mRNA | AK009379.1 | - | 2, 3 | 244 | ||
| AK019502.1 | - | 2, 3 | 262 | |||
| AK029897.1 | - | 2, 3 | 229 | |||
| AK051091.1 | - | 2, 3 | 220 | |||
| AK077335.1 | - | 2, 3 | 245 | |||
| AK161255.1 | - | 2, 3 | 219 | |||
| BC026535.1 | - | 2, 3 | 239 | |||
| BC037644.1 | - | 2, 3 | 241 | 5053565 | ||
| BQ931973.1 | - | 2, 3 | 265 | |||

Acad8
Genomic Map