siRNA Details
| Gene Symbol: Tmem206 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025864.3 | NP_080140.1 | 3 | 261 | ||
| GenBank mRNA | AK009526.1 | - | 3 | 262 | ||
| AK012107.1 | - | 3 | 279 | |||
| AK031270.1 | - | 3 | 295 | |||
| AK044482.1 | - | 3 | 262 | |||
| AK049641.1 | - | 3 | 298 | |||
| BC027151.1 | - | 3 | 255 | 4948749 | ||

Tmem206
Genomic Map