siRNA Details
| Gene Symbol: Efcab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025769.3 | NP_080045.1 | 3 | 415 | ||
| GenBank mRNA | AK015426.1 | - | 397 | |||
| AK015866.1 | - | 3 | 435 | |||
| AK017267.1 | - | 3 | 416 | |||
| AK038735.1 | - | 3 | 425 | |||
| AK042748.1 | - | 3 | 457 | |||
| AK043519.1 | - | 3 | 387 | |||
| AK077722.1 | - | 3 | 418 | |||
| BC085167.1 | - | 3 | 307 | 30538702 | ||

Efcab1
Genomic Map