siRNA Details
| Gene Symbol: Bcs1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012324.1 | - | 8 | 1393 | ||
| AK078925.1 | - | 8 | 1405 | |||
| AK079385.1 | - | 8 | 1401 | |||
| AK212348.1 | - | 1 | 170 | |||
| BC019781.1 | - | 8 | 1293 | 4036047 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bcs1l| Gene Symbol: Bcs1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012324.1 | - | 8 | 1393 | ||
| AK078925.1 | - | 8 | 1405 | |||
| AK079385.1 | - | 8 | 1401 | |||
| AK212348.1 | - | 1 | 170 | |||
| BC019781.1 | - | 8 | 1293 | 4036047 | ||