siRNA Details
| Gene Symbol: Nacc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025788.3 | NP_080064.3 | 6 | 2537 | ||
| GenBank mRNA | AK012935.1 | - | 6 | 2520 | ||
| BC048157.1 | - | 6 | 2510 | 5697941 | ||
| BC052706.1 | - | 6 | 2520 | 5702150 | ||
| BC054800.1 | - | 6 | 2537 | 6850337 | ||

Nacc1
Genomic Map