siRNA Details
| Gene Symbol: Plin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025836.3 | NP_080112.1 | 8 | 1625 | ||
| GenBank mRNA | AK003971.1 | - | 1 | 477 | ||
| AK004970.1 | - | 8 | 1627 | |||
| AK012517.1 | - | 8 | 1625 | |||
| AK132313.1 | - | 1626 | ||||
| AK167214.1 | - | 8 | 1625 | |||

Plin3
Genomic Map