siRNA Details
| Gene Symbol: Kdelr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025841.4 | NP_080117.1 | 5 | 1669 | ||
| GenBank mRNA | AK003515.1 | - | 5 | 1664 | ||
| AK150402.1 | - | 5 | 1664 | |||
| BC007146.1 | - | 5 | 1651 | 3496891 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kdelr2| Gene Symbol: Kdelr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025841.4 | NP_080117.1 | 5 | 1669 | ||
| GenBank mRNA | AK003515.1 | - | 5 | 1664 | ||
| AK150402.1 | - | 5 | 1664 | |||
| BC007146.1 | - | 5 | 1651 | 3496891 | ||