siRNA Details
| Gene Symbol: Vps28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF373710.1 | - | 10 | 885 | ||
| AK003572.2 | - | 10 | 915 | |||
| AK003699.1 | - | 10 | 839 | |||
| AK013852.1 | - | 10 | 836 | |||
| BC013535.1 | - | 10 | 848 | 4239189 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vps28| Gene Symbol: Vps28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF373710.1 | - | 10 | 885 | ||
| AK003572.2 | - | 10 | 915 | |||
| AK003699.1 | - | 10 | 839 | |||
| AK013852.1 | - | 10 | 836 | |||
| BC013535.1 | - | 10 | 848 | 4239189 | ||