siRNA Details
| Gene Symbol: Trim59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC006700.1 | - | 3 | 704 | 3501285 | |
| BC025430.1 | - | 3 | 719 | 4017983 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim59| Gene Symbol: Trim59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC006700.1 | - | 3 | 704 | 3501285 | |
| BC025430.1 | - | 3 | 719 | 4017983 | ||