siRNA Details
| Gene Symbol: Tmem206 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025864.3 | NP_080140.1 | 8 | 2127 | ||
| GenBank mRNA | AK009526.1 | - | 8 | 2127 | ||
| AK044482.1 | - | 8 | 2128 | |||
| AK049641.1 | - | 8 | 2164 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem206| Gene Symbol: Tmem206 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025864.3 | NP_080140.1 | 8 | 2127 | ||
| GenBank mRNA | AK009526.1 | - | 8 | 2127 | ||
| AK044482.1 | - | 8 | 2128 | |||
| AK049641.1 | - | 8 | 2164 | |||