siRNA Details
| Gene Symbol: Cdca7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025866.3 | NP_080142.1 | 9 | 1903 | ||
| GenBank mRNA | AK011289.1 | - | 9 | 1904 | ||
| AK028671.1 | - | 9 | 1915 | |||
| AK075882.1 | - | 9 | 1868 | |||
| AK153943.1 | - | 9 | 1882 | |||
| AK167734.1 | - | 9 | 1904 | |||
| BC066169.1 | - | 9 | 1911 | 30474357 | ||

Cdca7
Genomic Map