siRNA Details
Gene Symbol: Pole4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025882.3 | NP_080158.1 | 1 | 220 | ||
GenBank mRNA | AK010135.1 | - | 1 | 215 | ||
AK010300.1 | - | 1 | 221 | |||
AK011595.1 | - | 1 | 232 | |||
AK145365.1 | - | 1 | 226 | |||
AK160582.1 | - | 1 | 232 | |||
AK167631.1 | - | 1 | 210 | |||
BC023189.1 | - | 1 | 198 | 4952949 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
