siRNA Details
| Gene Symbol: Napa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025898.3 | NP_080174.1 | 1, 2 | 142 | ||
| GenBank mRNA | AK005372.1 | - | 1, 2 | 144 | ||
| AK013895.1 | - | 1, 2 | 161 | |||
| AK050623.1 | - | 1, 2 | 206 | |||
| AK136689.1 | - | 1, 2 | 145 | |||
| AK171998.1 | - | 1, 2 | 206 | |||
| BC004804.1 | - | 1, 2 | 173 | 3584869 | ||

Napa
Genomic Map