siRNA Details
| Gene Symbol: Ifrd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025903.2 | NP_080179.1 | 3 | 389 | ||
| GenBank mRNA | AK007687.1 | - | 3 | 390 | ||
| AK149496.1 | - | 3 | 370 | |||
| AK156625.1 | - | 3 | 383 | |||
| AK165967.1 | - | 3 | 382 | |||
| AK166130.1 | - | 3 | 457 | |||
| AK166258.1 | - | 3 | 347 | |||
| AY298860.1 | - | 3 | 218 | |||
| BC023486.1 | - | 3 | 337 | |||
| BC037434.1 | - | 3 | 345 | 2651172 | ||

Ifrd2
Genomic Map