siRNA Details
| Gene Symbol: Ttc23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168475.1 | NP_001161947.1 | 11 | 1915 | ||
| NM_001168476.1 | NP_001161948.1 | 10 | 1715 | |||
| NM_025905.3 | NP_080181.2 | 12 | 2041 | |||
| GenBank mRNA | AK005431.1 | - | 10 | 1676 | ||
| AK141754.1 | - | 11 | 1902 | |||
| BC038179.1 | - | 12 | 2012 | 4485073 | ||

Ttc23
Genomic Map