siRNA Details
| Gene Symbol: Ccdc91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025911.2 | NP_080187.2 | 4, 5 | 475 | ||
| GenBank mRNA | AK007017.1 | - | 3, 4 | 399 | ||
| AK007917.1 | - | 4, 5 | 476 | |||
| BC006734.1 | - | 4, 5 | 380 | 3156328 | ||
| BC049073.1 | - | 4, 5 | 444 | 5707404 | ||

Ccdc91
Genomic Map