siRNA Details
| Gene Symbol: Ccdc43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025918.3 | NP_080194.1 | 2 | 253 | ||
| GenBank mRNA | AK009319.1 | - | 2 | 249 | ||
| AK011254.1 | - | 2 | 254 | |||
| AK028834.1 | - | 2 | 264 | |||
| AK161543.1 | - | 2 | 264 | |||
| AK166820.1 | - | 2 | 249 | |||
| BC058520.1 | - | 2 | 262 | 6819447 | ||
| BC061076.1 | - | 2 | 262 | 6594359 | ||

Ccdc43
Genomic Map