siRNA Details
| Gene Symbol: 2610002M06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025921.3 | NP_080197.2 | 2, 3 | 291 | ||
| GenBank mRNA | AK011297.1 | - | 2, 3 | 239 | ||
| AK019383.1 | - | 2, 3 | 112 | |||
| BC016070.1 | - | 2, 3 | 230 | 4481241 | ||

2610002M06Rik
Genomic Map