siRNA Details
| Gene Symbol: Ift27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025931.2 | NP_080207.1 | 4 | 518 | ||
| GenBank mRNA | AK011196.1 | - | 4 | 516 | ||
| AK044278.1 | - | 1 | 956 | |||
| BC017514.1 | - | 4 | 518 | 4502942 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ift27| Gene Symbol: Ift27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025931.2 | NP_080207.1 | 4 | 518 | ||
| GenBank mRNA | AK011196.1 | - | 4 | 516 | ||
| AK044278.1 | - | 1 | 956 | |||
| BC017514.1 | - | 4 | 518 | 4502942 | ||