siRNA Details
| Gene Symbol: Lsm14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025948.2 | NP_080224.1 | 4, 5 | 608 | ||
| GenBank mRNA | AK003611.1 | - | 2, 3 | 168 | ||
| AK012276.1 | - | 3, 4 | 371 | |||
| AK186175.1 | - | 1, 2 | 128 | |||
| BC031521.1 | - | 4, 5 | 608 | 2655161 | ||

Lsm14a
Genomic Map